fac-Tris(2-phenylpyridine)iridium 98%

Supplier: Ambeed
Warning

Synonyms: fac-Ir(ppy)3

A206970-100MG A206970-250MG A206970-1G A206970-5G
103817-750EA 282 USD
103817-750 103848-430 76785-540 76785-542 77303-014 77303-016
fac-Tris(2-phenylpyridine)iridium 98%
fac-Tris(2-phenylpyridine)iridium

Precaución: for Research Use Only

Formula: C₃₃H₂₄IrN₃
MW: 654.78 g/mol
Melting Pt: 451 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12022527
CAS Number: 94928-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR