Ezetimibe 98%

Supplier: Ambeed
Danger

Synonyms: (3R,4S)-1-(4-Fluorophenyl)-3-((S)-3-(4-fluorophenyl)-3-hydroxypropyl)-4-(4-hydroxyphenyl)azetidin-2-one

A124629-100G A124629-100MG A124629-10G A124629-250MG A124629-25G A124629-50MG A124629-5G A124629-1G A124629-10MG
103800-560EA 24 USD
103800-560 76996-020 76996-022 76996-024 76996-026 76996-028 76996-030 77298-012 77312-696 77670-768
Ezetimibe 98%
Ezetimibe

Precaución: for Research Use Only

Formula: C₂₄H₂₁F₂NO₃
MW: 409.43 g/mol
Melting Pt: 164…166 °C
Flash Pt: 349.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00937872
CAS Number: 163222-33-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR