exo-Bicyclo[2.2.1]heptan-2-amine hydrochloride 97%

Supplier: Ambeed
Warning

A164431-250MG A164431-1G A164431-5G
103805-860EA 196 USD
103805-860 103844-118 77236-006 77300-714 77300-716
exo-Bicyclo[2.2.1]heptan-2-amine hydrochloride 97%
exo-Bicyclo[2.2.1]heptan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClN
MW: 147.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22124938
CAS Number: 39245-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR