Trimethylsilylacetylene 98%

Supplier: Ambeed
Danger

A407730-10G A407730-25G A407730-5G
103838-198EA 14 USD
103838-198 77155-652 77307-108 77307-110
Trimethylsilylacetylene 98%
Trimethylsilylacetylene

Precaución: for Research Use Only

Formula: C₅H₁₀Si
Boiling Pt: 52…53 °C (760 mm Hg)
Flash Pt: –34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008569
CAS Number: 1066-54-2
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR