(Triisopropylsilyl)acetylene 98%

Supplier: Ambeed
Danger

Synonyms: Ethynyltriisopropylsilane

A554152-10G A554152-25G A554152-5G A554152-1G
103828-704EA 9 USD
103828-704 103839-836 76790-268 76843-918 76843-920 77307-796
(Triisopropylsilyl)acetylene 98%
(Triisopropylsilyl)acetylene

Precaución: for Research Use Only

Formula: C₁₁H₂₂Si
MW: 182.38 g/mol
Boiling Pt: 50…52 °C at 0.6 mmHg
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075452
CAS Number: 89343-06-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR