(Ethyl)triphenylphosphonium bromide 98%
Supplier: Ambeed
|
Warning
|
A194526-100G
A194526-10G
A194526-500G
A194526-1000G
A194526-25G
103809-300EA
44
USD
103809-300
76800-106
76800-108
76800-110
77085-190
77085-192
(Ethyl)triphenylphosphonium bromide 98%
(Ethyl)triphenylphosphonium bromide
Precaución: for Research Use Only
Formula:
C₂₀H₂₀BrP Melting Pt: 203…205 °C Flash Pt: 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011838 CAS Number: 1530-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...