Ethyl undecanoate 98%

Supplier: Ambeed
Warning

A307632-100G A307632-10G A307632-500G A307632-25G
103835-688EA 6 USD
103835-688 76802-688 76802-690 76802-692 76838-434
Ethyl undecanoate 98%
Ethyl undecanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₆O₂
Boiling Pt: 105 °C (760 mm Hg)
Melting Pt: …15 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008958
CAS Number: 627-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR