Ethyl thiophene-3-carboxylate 95%

Supplier: Ambeed
Warning

A107321-25G A107321-1G A107321-5G A107321-100G A107321-10G
103797-170EA 10 USD
103797-170 103822-938 76765-562 77283-578 77309-784 77623-894 77623-896
Ethyl thiophene-3-carboxylate 95%
Ethyl thiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈O₂S
Boiling Pt: 211.7 °C (760 mm Hg)
Flash Pt: 103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179226
CAS Number: 5751-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR