Ethyl thiazole-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A214320-1G
A214320-25G
A214320-5G
A214320-10G
103818-390EA
9
USD
103818-390
103849-096
77087-612
77087-614
77087-616
77112-534
Ethyl thiazole-4-carboxylate 97%
Ethyl thiazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₇NO₂S Melting Pt: 51…55 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03788566 CAS Number: 14527-43-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...