Ethyl Quinuclidine-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl quinuclidine-4-carboxylate

A268799-1G A268799-5G
103812-934EA 21 USD
103812-934 103852-836 77305-014 77305-016
Ethyl Quinuclidine-4-carboxylate 98%
Ethyl Quinuclidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₂
MW: 183.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19686937
CAS Number: 22766-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR