Ethyl quinoline-6-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A228401-250MG
A228401-1G
103819-396EA
18
USD
103819-396
103850-304
76722-188
77089-094
Ethyl quinoline-6-carboxylate 97%
Ethyl quinoline-6-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₁NO₂ Boiling Pt: 328.3 °C (760 mm Hg) Flash Pt: 152.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047612 CAS Number: 73987-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...