Ethyl quinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

A210849-1G A210849-100MG A210849-250MG A210849-5G
103818-048EA 32 USD
103818-048 77087-126 77097-664 77097-666 77097-668
Ethyl quinoline-3-carboxylate 95%
Ethyl quinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂
MW: 201.22 g/mol
Boiling Pt: 309.2 °C (760 mm Hg)
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00957421
CAS Number: 50741-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR