Ethyl-3-piperidinecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl nipecotate, Ethyl 3-Piperidinecarboxylate, Ethyl piperidine-3-carboxylate

A242579-5G A242579-100G A242579-10G A242579-500G A242579-25G
103851-292EA 5 USD
103851-292 77090-474 77193-394 77193-396 77193-398 77304-246
Ethyl-3-piperidinecarboxylate 95%
Ethyl 3-piperidinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
Boiling Pt: 238.6 °C (760 mm Hg)
Flash Pt: 90.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005991
CAS Number: 5006-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR