Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate 97%

Supplier: Ambeed
Warning

A543210-25G A543210-5G A543210-100G A543210-10G A543210-500G
103839-768EA 5 USD
103839-768 76843-820 76843-822 77253-718 77253-720 77253-722
Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate 97%
Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate

Precaución: for Research Use Only

Formula: C₁₈H₂₁O₃P
MW: 316.3300 g/mol
Boiling Pt: 456 °C (760 mm Hg)
Flash Pt: 184 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08692488
CAS Number: 84434-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR