Ethyl-5-oxazolecarboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl oxazole-5-carboxylate
A127831-100G
A127831-10G
A127831-1G
A127831-25G
A127831-5G
103801-022EA
9
USD
103801-022
77009-762
77009-764
77313-290
77313-292
77313-294
Ethyl-5-oxazolecarboxylate 97%
Ethyl 5-oxazolecarboxylate
Precaución: for Research Use Only
Formula:
C₆H₇NO₃ MW: 141.13 g/mol Boiling Pt: 202 °C (760 mm Hg) Flash Pt: 79.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114930 CAS Number: 118994-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...