Ethyl-4-oxazolecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl oxazole-4-carboxylate

A110968-100G A110968-10G A110968-250MG A110968-25G A110968-1G A110968-5G
103797-766EA 6 USD
103797-766 103823-574 77160-730 77160-732 77160-734 77160-736 77310-378 77310-380
Ethyl-4-oxazolecarboxylate 97%
Ethyl 4-oxazolecarboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MW: 141.1300 g/mol
Boiling Pt: 202 °C (760 mm Hg)
Melting Pt: 48 °C
Flash Pt: 75.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04114940
CAS Number: 23012-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR