Ethyl-2-oxazolecarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl oxazole-2-carboxylate
A112697-250MG
A112697-1G
103798-078EA
23
USD
103798-078
103823-920
77160-830
77310-730
Ethyl-2-oxazolecarboxylate 95%
Ethyl 2-oxazolecarboxylate
Precaución: for Research Use Only
Formula:
C₆H₇NO₃ MW: 141.1300 g/mol Boiling Pt: 194 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD04114939 CAS Number: 33036-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...