Ethyl nitroacetate 99%

Supplier: Ambeed
Warning

Synonyms: Nitroethansäureethylester, Nitroacetic ethyl ester

A421737-25G A421737-5G A421737-100G A421737-10G A421737-1G A421737-500G
103838-454EA 6 USD
103838-454 76842-068 76842-070 77291-430 77291-432 77291-434 77291-436
Ethyl nitroacetate 99%
Ethyl nitroacetate

Precaución: for Research Use Only

Formula: C₄H₇NO₄
Boiling Pt: 208.6 °C (760 mm Hg)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007403
CAS Number: 626-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR