1-Boc-isonipecotic acid ethyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl N-BOC-4-piperidinecarboxylate, Ethyl N-Boc-piperidine-4-carboxylate

A119165-1000G A119165-100G A119165-10G A119165-1G A119165-25G A119165-500G A119165-5G
77149-100EA 155 USD
77149-100 77149-102 77149-104 77149-106 77149-108 77149-110 77297-662
1-Boc-isonipecotic acid ethyl ester 97%
1-Boc-isonipecotic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
Boiling Pt: 120…135 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01763998
CAS Number: 142851-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR