Ethyl-O-Mesitylsulfonylacetohydroxamate 98%

Supplier: Ambeed
Danger

Synonyms: O-Mesitylsulphonylacetohydroxamic acid Ethyl Ester, Ethyl N-(mesitylsulphonyl)oxyacetimidate

A110865-1G A110865-25G
103797-742EA 6 USD
103797-742 103823-550 77284-056 77565-948
Ethyl-O-Mesitylsulfonylacetohydroxamate 98%
Ethyl-O-Mesitylsulfonylacetohydroxamate

Precaución: For research use only.

Formula: C₁₃H₁₉NO₄S
Boiling Pt: 381.7 °C (760 mm Hg)
Melting Pt: 56…58 °C
Flash Pt: 184.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00009244
CAS Number: 38202-27-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR