Ethyl isoxazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A224155-100MG A224155-250MG
103819-088EA 25 USD
103819-088 103849-978 76814-982 76814-984
Ethyl isoxazole-5-carboxylate 95%
Ethyl isoxazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MW: 141.13 g/mol
Boiling Pt: 219.5±13.0 °C (760 mm Hg)
Flash Pt: 86.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18830699
CAS Number: 173850-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR