Ethyl indoline-6-carboxylate 97%

Supplier: Ambeed
Warning

A122531-250MG A122531-1G A122531-5G
103800-234EA 97 USD
103800-234 76822-518 77297-868 77297-870
Ethyl indoline-6-carboxylate 97%
Ethyl indoline-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09954776
CAS Number: 350683-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR