Ethyl indole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester

A362351-25G A362351-100G A362351-10G A362351-500G A362351-5G A362351-1000G
103837-332EA 10 USD
103837-332 76840-546 77278-346 77278-348 77278-350 77306-738 77748-721
Ethyl indole-2-carboxylate 98%
Ethyl indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
Boiling Pt: 342.4 °C (760 mm Hg)
Melting Pt: 122…125 °C
Flash Pt: 160.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00005609
CAS Number: 3770-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR