Ethyl-4-hydroxy-3-methoxycinnamate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3-methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-hydroxy-3-methoxyphenyl)acrylate, Ethyl Ferulate, Ferulic Acid Ethyl Ester

A172065-100G A172065-10G A172065-500G A172065-25G A172065-5G
103844-974EA 6 USD
103844-974 76720-542 76720-544 76720-546 77082-152 77301-182
Ethyl-4-hydroxy-3-methoxycinnamate 98%
Ethyl-4-hydroxy-3-methoxycinnamate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Boiling Pt: 382.3 °C (760 mm Hg)
Melting Pt: 63…65 °C
Flash Pt: 132.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00009190
CAS Number: 4046-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR