Ethyl diethoxyacetate 97%

Supplier: Ambeed
Warning

Synonyms: Diethoxyacetic ethyl ester

A227247-100G A227247-10G A227247-1G A227247-500G A227247-25G A227247-5G
103850-214EA 6 USD
103850-214 76771-028 76771-030 76771-032 76771-034 77088-978 77088-980
Ethyl diethoxyacetate 97%
Ethyl diethoxyacetate

Precaución: for Research Use Only

Formula: C₈H₁₆O₄
Boiling Pt: 199 °C (760 mm Hg)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00009231
CAS Number: 6065-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR