Ethyl benzo[d]isoxazole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1,2-Benzisoxazole-3-carboxylic acid ethyl ester, 3-Ethoxycarbonyl-1,2-benzisoxazole, Ethyl 1,2-Benzisoxazole-3-carboxylate

A125572-100MG A125572-250MG
103800-698EA 40 USD
103800-698 77026-758 77026-760
Ethyl benzo[d]isoxazole-3-carboxylate 97%
Ethyl benzo[d]isoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.1834 g/mol
Boiling Pt: 307 °C (760 mm Hg)
Melting Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD06659787
CAS Number: 57764-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR