Ethyl 7-chloro-1H-indole-2-carboxylate 95%

Supplier: Ambeed
Warning

A269311-100MG A269311-1G A269311-5G
103812-960EA 22 USD
103812-960 103852-866 76816-164 77092-628 77092-630
Ethyl 7-chloro-1H-indole-2-carboxylate 95%
Ethyl 7-chloro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClNO₂
MW: 223.66 g/mol
Boiling Pt: 375.0±22.0 °C (760 mm Hg)
Melting Pt: 105 °C
Flash Pt: 180.6 °C
Storage Temperature: Ambient
MDL Number: MFCD14715880
CAS Number: 43142-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR