Ethyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-3-(ethoxycarbonyl)imidazo[1,2-a]pyridine

A112494-1G A112494-5G
103798-020EA 27 USD
103798-020 103823-856 77310-664 77310-666
Ethyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate 97%
Ethyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11045695
CAS Number: 1134327-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR