Ethyl-7-aminoheptanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 7-aminoheptanoate hydrochloride, 7-Amino-heptanoic acid ethyl ester hydrochloride

A338212-25G A338212-100G A338212-10G A338212-1G
103825-770EA 9 USD
103825-770 103836-740 76839-764 77003-102 77003-104 77306-510
Ethyl-7-aminoheptanoate hydrochloride 97%
Ethyl-7-aminoheptanoate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₀ClNO₂
MW: 209.7100 g/mol
Boiling Pt: 269.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09743977
CAS Number: 29840-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR