Ethyl 6-oxo-3,6-dihydropyrimidine-4-carboxylate 98%

Supplier: Ambeed
Warning

A205846-250MG A205846-1G
103817-668EA 289 USD
103817-668 103848-334 77030-288 77302-970
Ethyl 6-oxo-3,6-dihydropyrimidine-4-carboxylate 98%
Ethyl 6-oxo-3,6-dihydropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113094
CAS Number: 223788-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR