Ethyl-6-nitroimidazo[1,2-a]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-nitroimidazo[1,2-a]pyridine-2-carboxylate

A126945-1G A126945-5G
103800-880EA 31 USD
103800-880 77298-160 77298-162
Ethyl-6-nitroimidazo[1,2-a]pyridine-3-carboxylate 97%
Ethyl 6-nitroimidazo[1,2-a]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉N₃O₄
MW: 235.2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864803
CAS Number: 38923-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR