Ethyl-6-iodoimidazo[1,2-a]pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-iodoimidazo[1,2-a]pyridine-2-carboxylate

A787813-250MG A787813-5G
103841-004EA 123 USD
103841-004 76791-586 76845-258
Ethyl-6-iodoimidazo[1,2-a]pyridine-2-carboxylate 98%
Ethyl 6-iodoimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉IN₂O₂
MW: 316.1000 g/mol
Melting Pt: 177…179 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02186151
CAS Number: 214958-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR