Ethyl 6-Hydroxyhexanoate 95%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxyhexanoic acid Ethyl ester, Ethyl 6-hydroxyhexanoate

A919967-100G A919967-1G A919967-5G
103841-402EA 10 USD
103841-402 76808-016 76808-018 76845-670
Ethyl 6-Hydroxyhexanoate 95%
Ethyl 6-Hydroxyhexanoate

Precaución: for Research Use Only

Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075601
CAS Number: 5299-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR