Ethyl 6-Chloropyridine-3-acetate 95%

Supplier: Ambeed
Warning

A110060-250MG A110060-1G A110060-5G A110060-10G
103797-610EA 62 USD
103797-610 103823-418 77093-960 77283-932 77283-934 77901-691
Ethyl 6-Chloropyridine-3-acetate 95%
Ethyl 6-Chloropyridine-3-acetate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
Boiling Pt: 285.8 °C (760 mm Hg)
Flash Pt: 126.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03092921
CAS Number: 197376-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR