Ethyl-6-chloroindole-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 6-chloroindole-2-carboxylate
A276223-5G
A276223-250MG
A276223-1G
103813-298EA
10
USD
103813-298
103853-232
77093-084
77276-872
77305-234
Ethyl-6-chloroindole-2-carboxylate 97%
Ethyl-6-chloroindole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₀ClNO₂ MW: 223.66 g/mol Boiling Pt: 375 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD03084731 CAS Number: 27034-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...