Ethyl-6-chloro-3-pyridazinecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-chloro-3-pyridazinecarboxylate, Ethyl 6-chloropyridazine-3-carboxylate, 6-chloro-3-pyridazinecarboxylic acid ethyl ester, 6-Chloro-pyridazine-3-carboxylic acid ethyl ester

A163183-10G A163183-250MG A163183-1G A163183-25G A163183-5G
103805-692EA 28 USD
103805-692 103843-952 76783-954 76783-956 77300-638 77300-640 77300-642
Ethyl-6-chloro-3-pyridazinecarboxylate 95%
Ethyl-6-chloro-3-pyridazinecarboxylate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.595 g/mol
Boiling Pt: 320 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09908168
CAS Number: 75680-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR