Ethyl 6-bromoquinoline-4-carboxylate 97%

Supplier: Ambeed
Warning

A290941-100MG A290941-250MG A290941-1G A290941-5G
103813-882EA 72 USD
103813-882 103835-026 76827-710 76827-712 76837-612 76837-614
Ethyl 6-bromoquinoline-4-carboxylate 97%
Ethyl 6-bromoquinoline-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₂
MW: 280.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135803
CAS Number: 220844-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR