Ethyl-6-bromopicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-bromopyridine-2-carboxylate

A151586-100G A151586-10G A151586-250MG A151586-1G A151586-25G A151586-5G
103804-276EA 9 USD
103804-276 103842-428 76997-352 76997-354 76997-356 77318-000 77318-002 77318-004
Ethyl-6-bromopicolinate 98%
Ethyl 6-bromopicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
Boiling Pt: 302.7 °C (760 mm Hg)
Melting Pt: 41…42 °C
Flash Pt: 136.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04116913
CAS Number: 21190-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR