Ethyl 6-bromoisoquinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

A100218-100MG A100218-1G
103795-302EA 119 USD
103795-302 103821-014 76982-120 77282-458
Ethyl 6-bromoisoquinoline-3-carboxylate 97%
Ethyl 6-bromoisoquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₂
MW: 280.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26793586
CAS Number: 882679-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR