Ethyl-6-aminopicolinate 98%

Supplier: Ambeed
Warning

Synonyms: 6-Aminopyridine-2-carboxylic acid ethyl ester, Ethyl 6-Aminopicolinate, Ethyl 6-Aminopyridine-2-carboxylate, 6-Aminopicolinic acid ethyl ester, Ethyl 6-aminopicolinate

A196215-100G A196215-10G A196215-250MG A196215-25G A196215-1G A196215-5G
103816-828EA 8 USD
103816-828 103847-440 76769-974 76769-976 76769-978 76769-980 77085-398 77085-400
Ethyl-6-aminopicolinate 98%
Ethyl-6-aminopicolinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
Boiling Pt: 329.6 °C (760 mm Hg)
Melting Pt: 56…58 °C
Flash Pt: 153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01009842
CAS Number: 69142-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR