Ethyl 6-aminonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-aminopyridine-5-carboxylate

A125257-100G A125257-10G A125257-250MG A125257-25G A125257-1G A125257-5G
103800-654EA 7 USD
103800-654 77189-854 77189-856 77189-858 77189-860 77312-804 77312-806
Ethyl 6-aminonicotinate 98%
Ethyl 6-aminonicotinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.177 g/mol
Boiling Pt: 314.7 °C (760 mm Hg)
Melting Pt: 145…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972544
CAS Number: 39658-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR