Ethyl-6-aminoimidazo[1,2-a]pyridine-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 6-aminoimidazo[1,2-a]pyridine-2-carboxylate
A159534-10G
A159534-250MG
A159534-1G
103805-266EA
36
USD
103805-266
103843-486
77096-104
77096-106
77300-408
Ethyl-6-aminoimidazo[1,2-a]pyridine-2-carboxylate 97%
Ethyl 6-aminoimidazo[1,2-a]pyridine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁N₃O₂ MW: 205.2132 g/mol Melting Pt: 169…171 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05864804 CAS Number: 158980-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...