Ethyl thiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-thiazolecarboxylate

A105981-250MG A105981-25G A105981-5G A105981-1G
103796-980EA 7 USD
103796-980 103822-730 76796-526 76796-528 77283-418 77309-592
Ethyl thiazole-5-carboxylate 98%
Ethyl thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MW: 157.1900 g/mol
Boiling Pt: 235.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06205069
CAS Number: 32955-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR