Ethyl 5-phenyl-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A104949-250MG A104949-5G A104949-1G
103796-804EA 13 USD
103796-804 103822-530 76765-446 77283-312 77309-392
Ethyl 5-phenyl-1H-pyrazole-3-carboxylate 97%
Ethyl 5-phenyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
Melting Pt: 140…141.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02130942
CAS Number: 5932-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR