Ethyl-5-nitroindole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-nitro-1H-indole-2-carboxylate

A219853-1G A219853-25G A219853-5G A219853-10G A219853-250MG
103818-822EA 13 USD
103818-822 103849-704 77088-276 77088-278 77088-280 77275-446 77275-448
Ethyl-5-nitroindole-2-carboxylate 98%
Ethyl-5-nitroindole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₄
MW: 234.211 g/mol
Boiling Pt: 429.5 °C (760 mm Hg)
Melting Pt: 220…225 °C
Flash Pt: 213.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216477
CAS Number: 16732-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR