Ethyl 5-methylpyrazole-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Methylpyrazole-3-carboxylic acid ethyl ester, Ethyl 5-methyl-1H-pyrazole-3-carboxylate
A267778-1G
A267778-5G
A267778-100G
A267778-10G
A267778-25G
A267778-500G
103812-882EA
9
USD
103812-882
103852-782
77092-530
77092-532
77113-846
77113-848
77304-990
77685-764
Ethyl 5-methylpyrazole-3-carboxylate 97%
Ethyl 5-methylpyrazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂O₂ Boiling Pt: 299.1 °C (760 mm Hg) Melting Pt: 80…84 °C Flash Pt: 134.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233455 CAS Number: 4027-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...