Ethyl 5-methylpyrazine-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A132279-100G
A132279-25G
A132279-1G
103801-708EA
9
USD
103801-708
76811-280
77298-550
77314-278
Ethyl 5-methylpyrazine-2-carboxylate 97%
Ethyl 5-methylpyrazine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₁₀N₂O₂ MW: 166.177 g/mol Boiling Pt: 251 °C (760 mm Hg) Flash Pt: 106 °C Storage Temperature: Ambient |
MDL Number:
MFCD05662506 CAS Number: 41110-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...