Ethyl-5-methylisoxazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-methylisoxazole-4-carboxylate

A147722-250MG A147722-25G A147722-1G
103803-816EA 14 USD
103803-816 103841-924 76972-918 76972-920 77299-596
Ethyl-5-methylisoxazole-4-carboxylate 97%
Ethyl 5-methylisoxazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₃
MW: 155.15 g/mol
Boiling Pt: 229.2 °C (760 mm Hg)
Flash Pt: 92.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01631119
CAS Number: 51135-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR