Ethyl-5-methylindole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-methylindole-2-carboxylate

A196233-100G A196233-10G A196233-1G A196233-25G A196233-5G
103816-832EA 33 USD
103816-832 103847-444 76721-284 76721-286 77085-404 77085-406 77085-408
Ethyl-5-methylindole-2-carboxylate 97%
Ethyl-5-methylindole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₂
Boiling Pt: 356.5 °C (760 mm Hg)
Melting Pt: 162…164 °C
Flash Pt: 169.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00022703
CAS Number: 16382-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR