Ethyl 5-methyl-1H-1,2,4-triazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A374528-1G A374528-5G A374528-100MG A374528-10G A374528-250MG A374528-25G
103826-552EA 36 USD
103826-552 103837-572 76840-848 76840-850 76978-736 76978-738 76978-740 77306-860
Ethyl 5-methyl-1H-1,2,4-triazole-3-carboxylate 97%
Ethyl 5-methyl-1H-1,2,4-triazole-3-carboxylate

Precaución: For research use only

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696752
CAS Number: 40253-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR